missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6-Fluorochromone-2-carboxylic Acid 97.0+%, TCI America™
$189.57 - $567.05
Chemical Identifiers
| CAS | 99199-59-4 |
|---|---|
| Molecular Formula | C10H5FO4 |
| Molecular Weight (g/mol) | 208.144 |
| MDL Number | MFCD03070543 |
| InChI Key | JZJYDFADRMBXAW-UHFFFAOYSA-N |
| Synonym | 6-fluorochromone-2-carboxylic acid, 6-fluoro-4-oxo-4h-chromene-2-carboxylic acid, 6-fluoro-4-oxo-4h-1-benzopyran-2-carboxylic acid, 6-fluoro-4-oxo-1-benzopyran-2-carboxylic acid, 6-fluoro-4-oxo-chromene-2-carboxylic acid, 4h-1-benzopyran-2-carboxylic acid, 6-fluoro-4-oxo, 6-fluorochromone-2-carboxylicacid, pubchem9710, acmc-209sbz |
| PubChem CID | 4096046 |
| IUPAC Name | 6-fluoro-4-oxochromene-2-carboxylic acid |
| SMILES | C1=CC2=C(C=C1F)C(=O)C=C(O2)C(=O)O |
Chemical Identifiers
| 99199-59-4 | |
| 208.144 | |
| JZJYDFADRMBXAW-UHFFFAOYSA-N | |
| 4096046 | |
| C1=CC2=C(C=C1F)C(=O)C=C(O2)C(=O)O |
| C10H5FO4 | |
| MFCD03070543 | |
| 6-fluorochromone-2-carboxylic acid, 6-fluoro-4-oxo-4h-chromene-2-carboxylic acid, 6-fluoro-4-oxo-4h-1-benzopyran-2-carboxylic acid, 6-fluoro-4-oxo-1-benzopyran-2-carboxylic acid, 6-fluoro-4-oxo-chromene-2-carboxylic acid, 4h-1-benzopyran-2-carboxylic acid, 6-fluoro-4-oxo, 6-fluorochromone-2-carboxylicacid, pubchem9710, acmc-209sbz | |
| 6-fluoro-4-oxochromene-2-carboxylic acid |
Specifications
| 99199-59-4 | |
| White-Yellow | |
| MFCD03070543 | |
| 6-fluorochromone-2-carboxylic acid, 6-fluoro-4-oxo-4h-chromene-2-carboxylic acid, 6-fluoro-4-oxo-4h-1-benzopyran-2-carboxylic acid, 6-fluoro-4-oxo-1-benzopyran-2-carboxylic acid, 6-fluoro-4-oxo-chromene-2-carboxylic acid, 4h-1-benzopyran-2-carboxylic acid, 6-fluoro-4-oxo, 6-fluorochromone-2-carboxylicacid, pubchem9710, acmc-209sbz | |
| C1=CC2=C(C=C1F)C(=O)C=C(O2)C(=O)O | |
| 208.144 | |
| 208.14 | |
| Crystalline Powder |
| 258°C | |
| C10H5FO4 | |
| 5 g | |
| JZJYDFADRMBXAW-UHFFFAOYSA-N | |
| 6-fluoro-4-oxochromene-2-carboxylic acid | |
| 4096046 | |
| ≥97.0% (HPLC,T) | |
| 6-Fluorochromone-2-carboxylic Acid |
Safety and Handling
TSCA : No