missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals 6-Fluoro-DL-tryptophan, 97%

Catalog No. p-2697685
Change view
Click to view available options
Quantity:
100 mg
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC146741000 100 mg Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC146741000 Supplier Thermo Scientific Chemicals Supplier No. 146741000

CAS: 7730-20-3 | C11H11FN2O2 | 222.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7730-20-3
Molecular Formula C11H11FN2O2
Molecular Weight (g/mol) 222.22
MDL Number MFCD00005653
InChI Key YMEXGEAJNZRQEH-UHFFFAOYSA-N
Synonym 6-fluoro-dl-tryptophan, 6-fluorotryptophan, 2-amino-3-6-fluoro-1h-indol-3-yl propanoic acid, dl-tryptophan, 6-fluoro, dl-6-fluorotryptophane, +--6-fluorotryptophan, tryptophan, 6-fluoro-, dl, 6-fluoro-d,l-tryptophan, tryptophan, 6-fluoro, 2-amino-3-6-fluoroindol-3-yl propanoic acid
PubChem CID 94937
IUPAC Name 2-amino-3-(6-fluoro-1H-indol-3-yl)propanoic acid
SMILES NC(CC1=CNC2=CC(F)=CC=C12)C(O)=O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 280°C to 285°C
Color Beige to White
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Quantity 100 mg
Formula Weight 222.22
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 6-Fluoro-DL-tryptophan
Show More Show Less
EINECSNumber 231-788-6

RUO – Research Use Only