accessibility menu, dialog, popup

UserName

NHONH2OHF

CAS RN 7730-20-3

IUPAC Name: 2-amino-3-(6-fluoro-1H-indol-3-yl)propanoic acid
Synonyms: 6-fluorotryptophan
Molecular Weight (g/mol): 222.22
Molecular Formula: C11H11FN2O2
InChi Key: YMEXGEAJNZRQEH-UHFFFAOYSA-N
SMILES: NC(CC1=CNC2=CC(F)=CC=C12)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results