missing translation for 'onlineSavingsMsg'
Learn More

6-Fluoro-4-chromanone 98.0+%, TCI America™

Catalog No. p-7129926
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F08311G 1 g
F08315G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F08311G Supplier TCI America Supplier No. F08311G

Chemical Identifiers

CAS 66892-34-0
Molecular Formula C9H7FO2
Molecular Weight (g/mol) 166.15
MDL Number MFCD00038654
InChI Key SWBBIJZMIGAZHW-UHFFFAOYSA-N
Synonym 6-fluorochroman-4-one, 6-fluoro-4-chromanone, 6-fluorochromanone, 6-fluoro-2,3-dihydro-4h-1-benzopyran-4-one, 6-fluorochromano-4-one, 6-fluoro-3,4-dihydro-2h-1-benzopyran-4-one, 6-fluoro-2,3-dihydro-4h-chromen-4-one, 6-fluoro-4-oxochroman, 6-fluoro-2,3-dihydro-1-benzopyran-4-one, 4h-1-benzopyran-4-one, 6-fluoro-2,3-dihydro
PubChem CID 2733262
IUPAC Name 6-fluoro-3,4-dihydro-2H-1-benzopyran-4-one
SMILES FC1=CC=C2OCCC(=O)C2=C1

Specifications

Melting Point 116°C
Quantity 1 g
Formula Weight 166.15
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 6-Fluoro-4-chromanone