accessibility menu, dialog, popup

UserName

FOO

CAS RN 66892-34-0

IUPAC Name: 6-fluoro-3,4-dihydro-2H-1-benzopyran-4-one
Synonyms: 6-fluoro-2,3-dihydro-1-benzopyran-4-one
Molecular Weight (g/mol): 166.15
Molecular Formula: C9H7FO2
InChi Key: SWBBIJZMIGAZHW-UHFFFAOYSA-N
SMILES: FC1=CC=C2OCCC(=O)C2=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results