missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6-Fluoro-2-pyridinecarboxylic Acid 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 138°C |
| Quantity | 5 g |
| Formula Weight | 141.10 |
| Percent Purity | ≥98.0% (T) |
| Grade | Not Available |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 6-Fluoro-2-pyridinecarboxylic Acid |
Safety and Handling
| TSCA | No |