accessibility menu, dialog, popup

UserName

HOOFN

CAS RN 402-69-7

IUPAC Name: 6-fluoropyridine-2-carboxylic acid
Synonyms: 6-fluoropyridine-2-carboxylic acid
Molecular Weight (g/mol): 141.1
Molecular Formula: C6H4FNO2
InChi Key: DIEMCUFYSOEIDU-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=CC(F)=N1
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)