missing translation for 'onlineSavingsMsg'
Learn More

6-Fluoro-2-pyridinecarboxylic Acid 98.0+%, TCI America™

Catalog No. F08371G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F08371G 1 g
F08375G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F08371G Supplier TCI America Supplier No. F08371G

Chemical Identifiers

CAS 402-69-7
Molecular Formula C6H4FNO2
Molecular Weight (g/mol) 141.10
MDL Number MFCD02181193
InChI Key DIEMCUFYSOEIDU-UHFFFAOYSA-N
Synonym 6-fluoropicolinic acid, 2-fluoropyridine-6-carboxylic acid, 6-fluoro-2-pyridinecarboxylic acid, 2-fluoro-6-pyridinecarboxylic acid, 2-fluoro-6-carboxypyridine, 6-fluoro-pyridine-2-carboxylic acid, 2-pyridinecarboxylic acid, 6-fluoro, 6-fluor-2-pyridincarbons, pubchem13570, 6-fluoro picolinic acid
PubChem CID 226027
IUPAC Name 6-fluoropyridine-2-carboxylic acid
SMILES OC(=O)C1=CC=CC(F)=N1

Specifications

Melting Point 138°C
Quantity 1 g
Formula Weight 141.10
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 6-Fluoro-2-pyridinecarboxylic Acid