missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6-(Dimethylamino)purine 98.0+%, TCI America™
$344.61 - $1151.25
Chemical Identifiers
| CAS | 938-55-6 |
|---|---|
| Molecular Formula | C7H9N5 |
| Molecular Weight (g/mol) | 163.184 |
| MDL Number | MFCD00005573 |
| InChI Key | BVIAOQMSVZHOJM-UHFFFAOYSA-N |
| PubChem CID | 3134 |
| ChEBI | CHEBI:60281 |
| IUPAC Name | N,N-dimethyl-7H-purin-6-amine |
| SMILES | CN(C)C1=NC=NC2=C1NC=N2 |
Chemical Identifiers
| 938-55-6 | |
| 163.184 | |
| BVIAOQMSVZHOJM-UHFFFAOYSA-N | |
| CHEBI:60281 | |
| CN(C)C1=NC=NC2=C1NC=N2 |
| C7H9N5 | |
| MFCD00005573 | |
| 3134 | |
| N,N-dimethyl-7H-purin-6-amine |
Specifications
| 938-55-6 | |
| White-Yellow | |
| MFCD00005573 | |
| 6-dimethylaminopurine, 6-dimethylamino purine, n,n-dimethyladenine, n6,n6-dimethyladenine, 6-dimethyladenine, dimethyladenine, n 6 ,n 6-dimethyladenine, n,n-dimethyl-9h-purin-6-amine, adenine, n,n-dimethyl, 6-dmap | |
| CN(C)C1=NC=NC2=C1NC=N2 | |
| 163.184 | |
| CHEBI:60281 | |
| ≥98.0% (HPLC,T) | |
| 6-(Dimethylamino)purine |
| 260°C | |
| C7H9N5 | |
| 1 g | |
| BVIAOQMSVZHOJM-UHFFFAOYSA-N | |
| N,N-dimethyl-7H-purin-6-amine | |
| 3134 | |
| 163.18 | |
| Crystalline Powder |
Safety and Handling
RTECSNumber : UO7440636
TSCA : No