accessibility menu, dialog, popup

UserName

CAS RN 938-55-6

IUPAC Name: N,N-dimethyl-7H-purin-6-amine
Synonyms: 6-(dimethylamino)-purine
Molecular Weight (g/mol): 163.18
Molecular Formula: C7H9N5
InChi Key: BVIAOQMSVZHOJM-UHFFFAOYSA-N
SMILES: CN(C)C1=C2NC=NC2=NC=N1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results