missing translation for 'onlineSavingsMsg'
Learn More

6'-(Diethylamino)-1',2'-benzofluoran 98.0+%, TCI America™

Catalog No. D320325G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D320325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D320325G Supplier TCI America Supplier No. D320325G
Only null left

Chemical Identifiers

CAS 26628-47-7
Molecular Formula C28H23NO3
Molecular Weight (g/mol) 421.50
MDL Number MFCD00480357
InChI Key HMNGPLGXLQFPFN-UHFFFAOYNA-N
PubChem CID 117321
IUPAC Name 8'-(diethylamino)-3H-10'-oxaspiro[2-benzofuran-1,5'-tetraphen]-3-one
SMILES CCN(CC)C1=CC=C2C(OC3=CC=C4C=CC=CC4=C3C22OC(=O)C3=CC=CC=C23)=C1

Specifications

Melting Point 221°C
Color Red-White
Quantity 25 g
Formula Weight 421.50
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 6′-(Diethylamino)-1′,2′-benzofluoran
EINECSNumber (5)-0660
TSCA Yes