accessibility menu, dialog, popup

UserName

H3CNCH3OOO

CAS RN 26628-47-7

IUPAC Name: 8'-(diethylamino)-3H-10'-oxaspiro[2-benzofuran-1,5'-tetraphen]-3-one
Synonyms: 8'-(diethylamino)-10'-oxaspiro[2-benzofuran-1,5'-tetraphen]-3-one
Molecular Weight (g/mol): 421.5
Molecular Formula: C28H23NO3
InChi Key: HMNGPLGXLQFPFN-UHFFFAOYNA-N
SMILES: CCN(CC)C1=CC=C2C(OC3=CC=C4C=CC=CC4=C3C22OC(=O)C3=CC=CC=C23)=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results