missing translation for 'onlineSavingsMsg'
Learn More

6-Cyano-7-nitro-1,4-dihydroquinoxaline-2,3-dione, 98%

Catalog No. AAJ63472MF
Change view
Click to view available options
Quantity:
10 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ63472MF 50 mg
AAJ63472MA 10 mg
AAJ63472MB 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ63472MF Supplier Thermo Scientific Chemicals Supplier No. J63472MF

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Potent, selective kainate/quisqualate (non-NMDA) receptor antagonist | CAS: 115066-14-3 | C9H4N4O4 | 232.155 g/mol

Potent, selective kainate/quisqualate (non-NMDA) receptor antagonist

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 115066-14-3
Molecular Formula C9H4N4O4
Molecular Weight (g/mol) 232.155
MDL Number MFCD00069232
InChI Key RPXVIAFEQBNEAX-UHFFFAOYSA-N
Synonym cnqx, 6-cyano-7-nitroquinoxaline-2,3-dione, unii-6ote87sccw, 7-nitro-2,3-dioxo-1,2,3,4-tetrahydroquinoxaline-6-carbonitrile, 6ote87sccw, 3h cnqx, 6-cyano-2,3-dihydroxy-7-nitroquinoxaline, 1,4-dihydro-2,3-dihydroxy-7-nitro-6-quinoxalinecarbonitrile, 6-quinoxalinecarbonitrile, 1,2,3,4-tetrahydro-7-nitro-2,3-dioxo, 2,3-dihydroxy-7-nitro-6-quinoxalinecarbonitrile
PubChem CID 3721046
ChEBI CHEBI:34468
IUPAC Name 7-nitro-2,3-dioxo-1,4-dihydroquinoxaline-6-carbonitrile
SMILES C1=C(C(=CC2=C1NC(=O)C(=O)N2)[N+](=O)[O-])C#N

Specifications

Melting Point >300°C
Color Yellow
Quantity 50 mg
Beilstein 5821072
Solubility Information Soluble in DMSO to 100mM
Formula Weight 232.16
Percent Purity ≥99%
Physical Form Solid
Chemical Name or Material 6-Cyano-7-nitro-1,4-dihydroquinoxaline-2,3-dione
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
RTECSNumber VD1520000
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only