missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6-Chloropyrazine-2-carboxylic Acid 96.0+%, TCI America™
$184.54 - $736.30
Chemical Identifiers
| CAS | 23688-89-3 |
|---|---|
| Molecular Formula | C5H3ClN2O2 |
| Molecular Weight (g/mol) | 158.541 |
| MDL Number | MFCD00837871 |
| InChI Key | KGGYMBKTQCLOTE-UHFFFAOYSA-N |
| Synonym | 6-chloro-pyrazine-2-carboxylic acid, 6-chloro-2-pyrazinecarboxylic acid, 2-carboxy-6-chloropyrazine, 2-chloro-6-carboxypyrazine, pyrazinecarboxylic acid, 6-chloro, 2-pyrazinecarboxylic acid, 6-chloro, 6-chloropyrazine-2-carboxylicacid, 6-chloro-pyrazine-2-carboxylicacid, pubchem16687, 6-chloro-2-pyrazinoic acid |
| PubChem CID | 14500875 |
| IUPAC Name | 6-chloropyrazine-2-carboxylic acid |
| SMILES | C1=C(N=C(C=N1)Cl)C(=O)O |
Chemical Identifiers
| 23688-89-3 | |
| 158.541 | |
| KGGYMBKTQCLOTE-UHFFFAOYSA-N | |
| 14500875 | |
| C1=C(N=C(C=N1)Cl)C(=O)O |
| C5H3ClN2O2 | |
| MFCD00837871 | |
| 6-chloro-pyrazine-2-carboxylic acid, 6-chloro-2-pyrazinecarboxylic acid, 2-carboxy-6-chloropyrazine, 2-chloro-6-carboxypyrazine, pyrazinecarboxylic acid, 6-chloro, 2-pyrazinecarboxylic acid, 6-chloro, 6-chloropyrazine-2-carboxylicacid, 6-chloro-pyrazine-2-carboxylicacid, pubchem16687, 6-chloro-2-pyrazinoic acid | |
| 6-chloropyrazine-2-carboxylic acid |
Specifications
| 23688-89-3 | |
| White-Yellow | |
| MFCD00837871 | |
| 6-chloro-pyrazine-2-carboxylic acid, 6-chloro-2-pyrazinecarboxylic acid, 2-carboxy-6-chloropyrazine, 2-chloro-6-carboxypyrazine, pyrazinecarboxylic acid, 6-chloro, 2-pyrazinecarboxylic acid, 6-chloro, 6-chloropyrazine-2-carboxylicacid, 6-chloro-pyrazine-2-carboxylicacid, pubchem16687, 6-chloro-2-pyrazinoic acid | |
| C1=C(N=C(C=N1)Cl)C(=O)O | |
| 158.541 | |
| 158.54 | |
| Crystalline Powder |
| 160°C | |
| C5H3ClN2O2 | |
| 1 g | |
| KGGYMBKTQCLOTE-UHFFFAOYSA-N | |
| 6-chloropyrazine-2-carboxylic acid | |
| 14500875 | |
| ≥96.0% (HPLC,T) | |
| 6-Chloropyrazine-2-carboxylic Acid |
Safety and Handling
TSCA : No