accessibility menu, dialog, popup

UserName

NNClOHO

CAS RN 23688-89-3

IUPAC Name: 6-chloropyrazine-2-carboxylic acid
Synonyms: 6-chloropyrazine-2-carboxylic acid
Molecular Weight (g/mol): 158.54
Molecular Formula: C5H3ClN2O2
InChi Key: KGGYMBKTQCLOTE-UHFFFAOYSA-N
SMILES: OC(=O)C1=CN=CC(Cl)=N1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results