missing translation for 'onlineSavingsMsg'
Learn More

6-Chloropyrazine-2-carboxylic Acid 96.0+%, TCI America™

Catalog No. C27255G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C27255G 5 g
C27251G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C27255G Supplier TCI America Supplier No. C27255G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 23688-89-3
Molecular Formula C5H3ClN2O2
Molecular Weight (g/mol) 158.541
MDL Number MFCD00837871
InChI Key KGGYMBKTQCLOTE-UHFFFAOYSA-N
Synonym 6-chloro-pyrazine-2-carboxylic acid, 6-chloro-2-pyrazinecarboxylic acid, 2-carboxy-6-chloropyrazine, 2-chloro-6-carboxypyrazine, pyrazinecarboxylic acid, 6-chloro, 2-pyrazinecarboxylic acid, 6-chloro, 6-chloropyrazine-2-carboxylicacid, 6-chloro-pyrazine-2-carboxylicacid, pubchem16687, 6-chloro-2-pyrazinoic acid
PubChem CID 14500875
IUPAC Name 6-chloropyrazine-2-carboxylic acid
SMILES C1=C(N=C(C=N1)Cl)C(=O)O

Specifications

Melting Point 160°C
Color White-Yellow
Quantity 5 g
Formula Weight 158.54
Percent Purity ≥96.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 6-Chloropyrazine-2-carboxylic Acid