missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6-Chloropurine 98.0+%, TCI America™
$136.08 - $300.11
Chemical Identifiers
| CAS | 87-42-3 |
|---|---|
| Molecular Formula | C5H3ClN4 |
| Molecular Weight (g/mol) | 154.557 |
| MDL Number | MFCD00075825 |
| InChI Key | ZKBQDFAWXLTYKS-UHFFFAOYSA-N |
| Synonym | 6-chloropurine, 6-chloro-9h-purine, 1h-purine, 6-chloro, 6-chloro-1h-purine, purine, 6-chloro, 6-cp, 6-chlorpurine, 6-chloro purine, 6-chloropurin, chloropurine, 6 |
| PubChem CID | 5359277 |
| IUPAC Name | 6-chloro-7H-purine |
| SMILES | C1=NC2=C(N1)C(=NC=N2)Cl |
Chemical Identifiers
| 87-42-3 | |
| 154.557 | |
| ZKBQDFAWXLTYKS-UHFFFAOYSA-N | |
| 5359277 | |
| C1=NC2=C(N1)C(=NC=N2)Cl |
| C5H3ClN4 | |
| MFCD00075825 | |
| 6-chloropurine, 6-chloro-9h-purine, 1h-purine, 6-chloro, 6-chloro-1h-purine, purine, 6-chloro, 6-cp, 6-chlorpurine, 6-chloro purine, 6-chloropurin, chloropurine, 6 | |
| 6-chloro-7H-purine |
Specifications
| 87-42-3 | |
| MFCD00075825 | |
| 6-chloropurine, 6-chloro-9h-purine, 1h-purine, 6-chloro, 6-chloro-1h-purine, purine, 6-chloro, 6-cp, 6-chlorpurine, 6-chloro purine, 6-chloropurin, chloropurine, 6 | |
| C1=NC2=C(N1)C(=NC=N2)Cl | |
| 154.557 | |
| 154.56 | |
| Crystalline Powder |
| C5H3ClN4 | |
| 5 g | |
| ZKBQDFAWXLTYKS-UHFFFAOYSA-N | |
| 6-chloro-7H-purine | |
| 5359277 | |
| ≥98.0% (T) | |
| 6-Chloropurine |
Safety and Handling
EINECSNumber : (5)-1130
RTECSNumber : UO7520000
TSCA : No