missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6-Chloropurine 98.0+%, TCI America™
Supplier: TCI America C027825G
Specifications
| 6-Chloropurine | |
| C5H3ClN4 | |
| 25 g | |
| ZKBQDFAWXLTYKS-UHFFFAOYSA-N | |
| 6-chloro-7H-purine | |
| 5359277 | |
| ≥98.0% (T) |
| 87-42-3 | |
| MFCD00075825 | |
| 6-chloropurine, 6-chloro-9h-purine, 1h-purine, 6-chloro, 6-chloro-1h-purine, purine, 6-chloro, 6-cp, 6-chlorpurine, 6-chloro purine, 6-chloropurin, chloropurine, 6 | |
| C1=NC2=C(N1)C(=NC=N2)Cl | |
| 154.557 | |
| 154.56 | |
| Crystalline Powder |
Chemical Identifiers
| 87-42-3 | |
| 154.557 | |
| ZKBQDFAWXLTYKS-UHFFFAOYSA-N | |
| 5359277 | |
| C1=NC2=C(N1)C(=NC=N2)Cl |
| C5H3ClN4 | |
| MFCD00075825 | |
| 6-chloropurine, 6-chloro-9h-purine, 1h-purine, 6-chloro, 6-chloro-1h-purine, purine, 6-chloro, 6-cp, 6-chlorpurine, 6-chloro purine, 6-chloropurin, chloropurine, 6 | |
| 6-chloro-7H-purine |
Safety and Handling
EINECSNumber : (5)-1130
RTECSNumber : UO7520000
TSCA : No