missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6-Chloropurine Riboside 97.0+%, TCI America™
$148.10 - $512.62
Chemical Identifiers
| CAS | 5399-87-1 |
|---|---|
| Molecular Formula | C10H11ClN4O4 |
| Molecular Weight (g/mol) | 286.67 |
| MDL Number | MFCD00005738 |
| InChI Key | XHRJGHCQQPETRH-UHFFFAOYNA-N |
| Synonym | 6-chloropurine riboside, chloropurine riboside, 2r,3r,4s,5r-2-6-chloro-9h-purin-9-yl-5-hydroxymethyl tetrahydrofuran-3,4-diol, 6-chloropurine ribonucleoside, 6-chloropurine-9-riboside, 6-chloro-9-ribofuranosyl-9h-purine, 9h-purine, 6-chloro-9-ribofuranosyl, 6-chloropurineriboside, 6-chloro-9-beta-d-ribofuranosyl-9h-purine, 6-chloronebularine |
| PubChem CID | 93003 |
| IUPAC Name | 2-(6-chloro-9H-purin-9-yl)-5-(hydroxymethyl)oxolane-3,4-diol |
| SMILES | OCC1OC(C(O)C1O)N1C=NC2=C(Cl)N=CN=C12 |
Chemical Identifiers
| 5399-87-1 | |
| 286.67 | |
| XHRJGHCQQPETRH-UHFFFAOYNA-N | |
| 93003 | |
| OCC1OC(C(O)C1O)N1C=NC2=C(Cl)N=CN=C12 |
| C10H11ClN4O4 | |
| MFCD00005738 | |
| 6-chloropurine riboside, chloropurine riboside, 2r,3r,4s,5r-2-6-chloro-9h-purin-9-yl-5-hydroxymethyl tetrahydrofuran-3,4-diol, 6-chloropurine ribonucleoside, 6-chloropurine-9-riboside, 6-chloro-9-ribofuranosyl-9h-purine, 9h-purine, 6-chloro-9-ribofuranosyl, 6-chloropurineriboside, 6-chloro-9-beta-d-ribofuranosyl-9h-purine, 6-chloronebularine | |
| 2-(6-chloro-9H-purin-9-yl)-5-(hydroxymethyl)oxolane-3,4-diol |
Specifications
| 5399-87-1 | |
| MFCD00005738 | |
| 6-chloropurine riboside, chloropurine riboside, 2r,3r,4s,5r-2-6-chloro-9h-purin-9-yl-5-hydroxymethyl tetrahydrofuran-3,4-diol, 6-chloropurine ribonucleoside, 6-chloropurine-9-riboside, 6-chloro-9-ribofuranosyl-9h-purine, 9h-purine, 6-chloro-9-ribofuranosyl, 6-chloropurineriboside, 6-chloro-9-beta-d-ribofuranosyl-9h-purine, 6-chloronebularine | |
| OCC1OC(C(O)C1O)N1C=NC2=C(Cl)N=CN=C12 | |
| 286.67 | |
| 286.67 | |
| Crystalline Powder |
| C10H11ClN4O4 | |
| 1 g | |
| XHRJGHCQQPETRH-UHFFFAOYNA-N | |
| 2-(6-chloro-9H-purin-9-yl)-5-(hydroxymethyl)oxolane-3,4-diol | |
| 93003 | |
| ≥97.0% (HPLC,T) | |
| 6-Chloropurine Riboside |
Safety and Handling
RTECSNumber : UO7520800
TSCA : No