missing translation for 'onlineSavingsMsg'
Learn More

6-Chloropurine Riboside 97.0+%, TCI America™

Catalog No. C22061G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C22061G 1 g
C22065G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C22061G Supplier TCI America Supplier No. C22061G

Chemical Identifiers

CAS 5399-87-1
Molecular Formula C10H11ClN4O4
Molecular Weight (g/mol) 286.67
MDL Number MFCD00005738
InChI Key XHRJGHCQQPETRH-UHFFFAOYNA-N
Synonym 6-chloropurine riboside, chloropurine riboside, 2r,3r,4s,5r-2-6-chloro-9h-purin-9-yl-5-hydroxymethyl tetrahydrofuran-3,4-diol, 6-chloropurine ribonucleoside, 6-chloropurine-9-riboside, 6-chloro-9-ribofuranosyl-9h-purine, 9h-purine, 6-chloro-9-ribofuranosyl, 6-chloropurineriboside, 6-chloro-9-beta-d-ribofuranosyl-9h-purine, 6-chloronebularine
PubChem CID 93003
IUPAC Name 2-(6-chloro-9H-purin-9-yl)-5-(hydroxymethyl)oxolane-3,4-diol
SMILES OCC1OC(C(O)C1O)N1C=NC2=C(Cl)N=CN=C12

Specifications

Quantity 1 g
Formula Weight 286.67
Percent Purity ≥97.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 6-Chloropurine Riboside
RTECSNumber UO7520800
TSCA No