missing translation for 'onlineSavingsMsg'
Learn More

6-Chloropurine riboside, 99+%

Catalog No. AC226450050
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC226450050 5 g Glass bottle
AC226450010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC226450050 Supplier Thermo Scientific Chemicals Supplier No. 226450050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 5399-87-1 | C10H11ClN4O4 | 286.67 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5399-87-1
Molecular Formula C10H11ClN4O4
Molecular Weight (g/mol) 286.67
MDL Number MFCD00005738
InChI Key XHRJGHCQQPETRH-UHFFFAOYNA-N
Synonym 6-chloropurine riboside, chloropurine riboside, 2r,3r,4s,5r-2-6-chloro-9h-purin-9-yl-5-hydroxymethyl tetrahydrofuran-3,4-diol, 6-chloropurine ribonucleoside, 6-chloropurine-9-riboside, 6-chloro-9-ribofuranosyl-9h-purine, 9h-purine, 6-chloro-9-ribofuranosyl, 6-chloropurineriboside, 6-chloro-9-beta-d-ribofuranosyl-9h-purine, 6-chloronebularine
PubChem CID 93003
IUPAC Name (2R,3R,4S,5R)-2-(6-chloropurin-9-yl)-5-(hydroxymethyl)oxolane-3,4-diol
SMILES OCC1OC(C(O)C1O)N1C=NC2=C(Cl)N=CN=C12

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point 161°C to 163°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 99% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Formula Weight 286.66
Percent Purity 99+%
Grade Pure
Physical Form Fine Crystalline Powder
Chemical Name or Material 6-Chloropurine riboside, Pure
Show More Show Less
EINECSNumber 226-438-4

RUO – Research Use Only