missing translation for 'onlineSavingsMsg'
Learn More

6-Chloronicotinamide, 98%

Catalog No. AAB2192203
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2192203 1 g
AAB2192206 5 g
AAB2192214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2192203 Supplier Thermo Scientific Chemicals Supplier No. B2192203

CAS: 6271-78-9 | C6H5ClN2O | 156.57 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6271-78-9
Molecular Formula C6H5ClN2O
Molecular Weight (g/mol) 156.57
MDL Number MFCD00006242
InChI Key ZIJAZUBWHAZHPL-UHFFFAOYSA-N
Synonym 6-chloronicotinamide, 3-pyridinecarboxamide, 6-chloro, nicotinamide, 6-chloro, 6-chloro-3-pyridinecarboxamide, 6-chloro-nicotinamide, acmc-1b4gd, 6-chloronicotinic acid amide, 3-carbamoyl-6-chloropyridine, ksc492o5t
PubChem CID 80456
IUPAC Name 6-chloropyridine-3-carboxamide
SMILES NC(=O)C1=CC=C(Cl)N=C1

Specifications

Melting Point 214°C to 216°C
Quantity 1 g
Beilstein 116043
Formula Weight 156.57
Percent Purity 98%
Chemical Name or Material 6-Chloronicotinamide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 228-456-8
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only