missing translation for 'onlineSavingsMsg'
Learn More

6-(chloromethyl)benzothiazole, 97%, Thermo Scientific™

Chemical Identifiers

CAS 149440-49-3
Molecular Formula C8H6ClNS
Molecular Weight (g/mol) 183.66
InChI Key WVJSICZOZHJCLJ-UHFFFAOYSA-N
Synonym 6-chloromethyl benzo d thiazole, 6-chloromethyl benzothiazole, benzothiazole, 6-chloromethyl, 6-chloromethyl-1,3-benzothiazole, acmc-20n5pd, 6-chloromethylbenzothiazole
PubChem CID 22905771
IUPAC Name 6-(chloromethyl)-1,3-benzothiazole
SMILES C1=CC2=C(C=C1CCl)SC=N2
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC441350010
SDS View Documents
Thermo Scientific Chemicals
441350010
1g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

149440-49-3
183.66
6-chloromethyl benzo d thiazole, 6-chloromethyl benzothiazole, benzothiazole, 6-chloromethyl, 6-chloromethyl-1,3-benzothiazole, acmc-20n5pd, 6-chloromethylbenzothiazole
6-(chloromethyl)-1,3-benzothiazole
C8H6ClNS
WVJSICZOZHJCLJ-UHFFFAOYSA-N
22905771
C1=CC2=C(C=C1CCl)SC=N2

Specifications

149440-49-3
100.0
96% min. (GC)
C8H6ClNS
6-chloromethyl benzo d thiazole, 6-chloromethyl benzothiazole, benzothiazole, 6-chloromethyl, 6-chloromethyl-1,3-benzothiazole, acmc-20n5pd, 6-chloromethylbenzothiazole
C1=CC2=C(C=C1CCl)SC=N2
183.66
183.66
6-(Chloromethyl)benzothiazole
96.0
Authentic
Glass Bottle
1g
WVJSICZOZHJCLJ-UHFFFAOYSA-N
6-(chloromethyl)-1,3-benzothiazole
22905771
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Harmful if swallowed.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only