accessibility menu, dialog, popup

UserName

CAS RN 149440-49-3

ClSN

CAS RN 149440-49-3

IUPAC Name: 6-(chloromethyl)-1,3-benzothiazole
Synonyms: 6-(chloromethyl)-1,3-benzothiazole
Molecular Weight (g/mol): 183.65
Molecular Formula: C8H6ClNS
InChi Key: WVJSICZOZHJCLJ-UHFFFAOYSA-N
SMILES: ClCC1=CC=C2N=CSC2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)