missing translation for 'onlineSavingsMsg'
Learn More

6-Chloroindole-2-carboxylic acid, 96%, Thermo Scientific Chemicals

Catalog No. AAH6686406
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6686406 5 g
AAH6686403 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6686406 Supplier Thermo Scientific Chemicals Supplier No. H6686406
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 16732-75-5
Molecular Formula C9H6ClNO2
Molecular Weight (g/mol) 195.602
MDL Number MFCD01863157
InChI Key BKPSJOSKWKTWAG-UHFFFAOYSA-N
Synonym 6-chloroindole-2-carboxylic acid, pubchem1671, acmc-209dwc, chembl23134, 6-chloro-2-indolecarboxylic acid, 6-chloro indole-2-carboxylic acid, 1h-indole-2-carboxylic acid, 6-chloro, 6-chloro-1h-indole-2-carboxylic acid; 6-chloro-1h-indole-2-carboxylic acid; 6-chloro-2-indolecarboxylic acid
PubChem CID 85584
IUPAC Name 6-chloro-1H-indole-2-carboxylic acid
SMILES C1=CC2=C(C=C1Cl)NC(=C2)C(=O)O

Specifications

Quantity 5 g
Beilstein 152660
Formula Weight 195.6
Percent Purity 96%
Physical Form Solid
Chemical Name or Material 6-Chloroindole-2-carboxylic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 240-799-5
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only