missing translation for 'onlineSavingsMsg'
Learn More

6-Chloroimidazo[1,2-b]pyridazine 98.0+%, TCI America™

Catalog No. C31275G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C31275G 5 g
C31271G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C31275G Supplier TCI America Supplier No. C31275G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 6775-78-6
Molecular Formula C6H4ClN3
Molecular Weight (g/mol) 153.569
MDL Number MFCD07778345
InChI Key MPZDNIJHHXRTIQ-UHFFFAOYSA-N
PubChem CID 138828
IUPAC Name 6-chloroimidazo[1,2-b]pyridazine
SMILES C1=CC(=NN2C1=NC=C2)Cl

Specifications

Melting Point 116°C
Quantity 5 g
Formula Weight 153.57
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 6-Chloroimidazo[1,2-b]pyridazine