accessibility menu, dialog, popup

UserName

CAS RN 6775-78-6

IUPAC Name: 6-chloroimidazo[1,2-b]pyridazine
Synonyms: 6-chloroimidazo[1,2-b]pyridazine
Molecular Weight (g/mol): 153.57
Molecular Formula: C6H4ClN3
InChi Key: MPZDNIJHHXRTIQ-UHFFFAOYSA-N
SMILES: ClC1=NN2C=CN=C2C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results