missing translation for 'onlineSavingsMsg'
Learn More

6-Chloroflavone 98.0+%, TCI America™

Catalog No. C30661G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C30661G 1 g
C30665G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C30661G Supplier TCI America Supplier No. C30661G
Only null left

Chemical Identifiers

CAS 10420-73-2
Molecular Formula C15H9ClO2
Molecular Weight (g/mol) 256.685
MDL Number MFCD00209560
InChI Key IFNDLWHUYFSXBK-UHFFFAOYSA-N
Synonym 6-chloroflavone, 6-chloro-2-phenyl-4h-chromen-4-one, 6-chloro-2-phenyl-chromen-4-one, 6-chlorflavone, maybridge4_000538, d02chg, 6-chloro-2-phenyl-4h-chromen-4-one #, 6-chloro-2-phenyl-4h-1-benzopyran-4-one
PubChem CID 248021
IUPAC Name 6-chloro-2-phenylchromen-4-one
SMILES C1=CC=C(C=C1)C2=CC(=O)C3=C(O2)C=CC(=C3)Cl

Specifications

Melting Point 185°C
Quantity 1 g
Formula Weight 256.69
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 6-Chloroflavone