accessibility menu, dialog, popup

UserName

CAS RN 10420-73-2

IUPAC Name: 6-chloro-2-phenyl-4H-chromen-4-one
Synonyms: 6-chloroflavone
Molecular Weight (g/mol): 256.69
Molecular Formula: C15H9ClO2
InChi Key: IFNDLWHUYFSXBK-UHFFFAOYSA-N
SMILES: ClC1=CC=C2OC(=CC(=O)C2=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results