missing translation for 'onlineSavingsMsg'
Learn More

6-Chlorochromone-2-carboxylic Acid 98.0+%, TCI America™

Catalog No. C31741G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C31741G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C31741G Supplier TCI America Supplier No. C31741G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 5006-45-1
Molecular Formula C10H5ClO4
Molecular Weight (g/mol) 224.596
MDL Number MFCD00847039
InChI Key HALQFUWRVXLBIS-UHFFFAOYSA-N
Synonym 6-Chloro-4-oxo-4H-1-benzopyran-2-carboxylic Acid
PubChem CID 21107
IUPAC Name 6-chloro-4-oxochromene-2-carboxylic acid
SMILES C1=CC2=C(C=C1Cl)C(=O)C=C(O2)C(=O)O

Specifications

Melting Point 269°C
Color White
Quantity 1 g
Formula Weight 224.60
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 6-Chlorochromone-2-carboxylic Acid
RTECSNumber DJ2260650
TSCA No