accessibility menu, dialog, popup

UserName

OOOClO

CAS RN 5006-45-1

IUPAC Name: 6-chloro-4-oxo-4H-chromene-2-carboxylate
Synonyms: 6-chloro-4-oxochromene-2-carboxylate
Molecular Weight (g/mol): 223.59
Molecular Formula: C10H4ClO4
InChi Key: HALQFUWRVXLBIS-UHFFFAOYSA-M
SMILES: [O-]C(=O)C1=CC(=O)C2=CC(Cl)=CC=C2O1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results