missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6-Chloro-7-iodo-7-deazapurine 95.0+%, TCI America™
$55.92 - $219.95
Chemical Identifiers
| CAS | 123148-78-7 |
|---|---|
| Molecular Formula | C6H3ClIN3 |
| Molecular Weight (g/mol) | 279.465 |
| MDL Number | MFCD09263258 |
| InChI Key | CBWBJFJMNBPWAL-UHFFFAOYSA-N |
| Synonym | 4-chloro-5-iodo-7h-pyrrolo 2,3-d pyrimidine, 6-chloro-7-iodo-7-deazapurine, 4-chloro-5-iodo-7h-pyrrol 2,3-d pyrimidine, 4-chloro-5-iodo-1h-pyrrolo 2,3-d pyrimidine, 4-chloro-5-iodopyrrolo 2,3-d pyrimidine, 7h-pyrrolo 2,3-d pyrimidine, 4-chloro-5-iodo, acmc-20agy2, amtns039, 6-chloro-7-iod-7-deazapurine |
| PubChem CID | 14809281 |
| IUPAC Name | 4-chloro-5-iodo-7H-pyrrolo[2,3-d]pyrimidine |
| SMILES | C1=C(C2=C(N1)N=CN=C2Cl)I |
Chemical Identifiers
| 123148-78-7 | |
| 279.465 | |
| CBWBJFJMNBPWAL-UHFFFAOYSA-N | |
| 14809281 | |
| C1=C(C2=C(N1)N=CN=C2Cl)I |
| C6H3ClIN3 | |
| MFCD09263258 | |
| 4-chloro-5-iodo-7h-pyrrolo 2,3-d pyrimidine, 6-chloro-7-iodo-7-deazapurine, 4-chloro-5-iodo-7h-pyrrol 2,3-d pyrimidine, 4-chloro-5-iodo-1h-pyrrolo 2,3-d pyrimidine, 4-chloro-5-iodopyrrolo 2,3-d pyrimidine, 7h-pyrrolo 2,3-d pyrimidine, 4-chloro-5-iodo, acmc-20agy2, amtns039, 6-chloro-7-iod-7-deazapurine | |
| 4-chloro-5-iodo-7H-pyrrolo[2,3-d]pyrimidine |
Specifications
| 123148-78-7 | |
| White-Yellow | |
| MFCD09263258 | |
| 4-chloro-5-iodo-7h-pyrrolo 2,3-d pyrimidine, 6-chloro-7-iodo-7-deazapurine, 4-chloro-5-iodo-7h-pyrrol 2,3-d pyrimidine, 4-chloro-5-iodo-1h-pyrrolo 2,3-d pyrimidine, 4-chloro-5-iodopyrrolo 2,3-d pyrimidine, 7h-pyrrolo 2,3-d pyrimidine, 4-chloro-5-iodo, acmc-20agy2, amtns039, 6-chloro-7-iod-7-deazapurine | |
| C1=C(C2=C(N1)N=CN=C2Cl)I | |
| 279.465 | |
| 279.47 | |
| Crystalline Powder |
| 202°C | |
| C6H3ClIN3 | |
| 1 g | |
| CBWBJFJMNBPWAL-UHFFFAOYSA-N | |
| 4-chloro-5-iodo-7H-pyrrolo[2,3-d]pyrimidine | |
| 14809281 | |
| ≥95.0% (GC,T) | |
| 6-Chloro-7-iodo-7-deazapurine |
Safety and Handling
TSCA : No