accessibility menu, dialog, popup

UserName

HNNNClI

CAS RN 123148-78-7

IUPAC Name: 4-chloro-5-iodo-7H-pyrrolo[2,3-d]pyrimidine
Synonyms: 4-chloro-5-iodo-7H-pyrrolo[2,3-d]pyrimidine
Molecular Weight (g/mol): 279.47
Molecular Formula: C6H3ClIN3
InChi Key: CBWBJFJMNBPWAL-UHFFFAOYSA-N
SMILES: ClC1=C2C(I)=CNC2=NC=N1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results