missing translation for 'onlineSavingsMsg'
Learn More

6-Chloro-7-azaindole, 95%

Catalog No. p-7057215
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6665503 1 g
AAH6665506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6665503 Supplier Thermo Scientific Chemicals Supplier No. H6665503
Only null left

CAS: 55052-27-2 | C7H5ClN2 | 152.58 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 55052-27-2
Molecular Formula C7H5ClN2
Molecular Weight (g/mol) 152.58
MDL Number MFCD06659666
InChI Key DSCOSIUAQUDGJM-UHFFFAOYSA-N
Synonym 6-chloro-1h-pyrrolo 2,3-b pyridine, 6-chloro-7-azaindole, 1h-pyrrolo 2,3-b pyridine, 6-chloro, 7-aza-6-chloroindole, pubchem18673, ksc494a8j, 6-chloranyl-1h-pyrrolo 2,3-b pyridine
PubChem CID 11344118
IUPAC Name 6-chloro-1H-pyrrolo[2,3-b]pyridine
SMILES ClC1=CC=C2C=CNC2=N1

Specifications

Melting Point 170°C to 175°C
Color Brown to White
Quantity 1 g
UN Number UN2811
Beilstein 907449
Formula Weight 152.58
Percent Purity 95%
Physical Form Powder
Chemical Name or Material 6-Chloro-7-azaindole
Hazard Category H302-H315-H318-H335
Hazard Statement GHS H Statement
H318-H302-H335-H315
Causes serious eye damage.
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only