accessibility menu, dialog, popup

UserName

NHNCl

CAS RN 55052-27-2

IUPAC Name: 6-chloro-1H-pyrrolo[2,3-b]pyridine
Synonyms: 6-chloro-1H-pyrrolo[2,3-b]pyridine
Molecular Weight (g/mol): 152.58
Molecular Formula: C7H5ClN2
InChi Key: DSCOSIUAQUDGJM-UHFFFAOYSA-N
SMILES: ClC1=CC=C2C=CNC2=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results