missing translation for 'onlineSavingsMsg'
Learn More

6-Chloro-5-nitro-2-picoline, 98%

Catalog No. p-3736002
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC360830010 1 g Glass bottle
AC360830050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC360830010 Supplier Thermo Scientific Chemicals Supplier No. 360830010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 56057-19-3 | C6H5ClN2O2 | 172.5 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 56057-19-3
Molecular Formula C6H5ClN2O2
Molecular Weight (g/mol) 172.5
MDL Number MFCD03085820
InChI Key UIEVSGOVFXWCIK-UHFFFAOYSA-N
Synonym 6-chloro-5-nitro-2-picoline, 2-chloro-3-nitro-6-methylpyridine, 2-chloro-3-nitro-6-picoline, 2-chloro-3-nitro-6-methyl pyridine, 2-chloro-6-methyl-3-nitro-pyridine, pyridine, 2-chloro-6-methyl-3-nitro, 2-chloro-6-methyl-3-nitropyridine?, pubchem6175, acmc-209lqq
PubChem CID 1268230
IUPAC Name 2-chloro-6-methyl-3-nitropyridine
SMILES CC1=NC(=C(C=C1)[N+](=O)[O-])Cl

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 70°C to 74°C
Color Brown
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 172.5
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 6-Chloro-5-nitro-2-picoline
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Harmful if swallowed.
May cause respiratory irritation.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only