missing translation for 'onlineSavingsMsg'
Learn More

6-Chloro-4-hydroxy-2-methylpyrimidine, 97%

Catalog No. AAH3347803
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3347803 1 g
AAH3347809 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3347803 Supplier Thermo Scientific Chemicals Supplier No. H3347803
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 17551-52-9 | C5H5ClN2O | 144.558 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 17551-52-9
Molecular Formula C5H5ClN2O
Molecular Weight (g/mol) 144.558
MDL Number MFCD00030773
InChI Key QNANRGHPXMUJQC-UHFFFAOYSA-N
Synonym 6-chloro-2-methyl-4-pyrimidinol, 6-chloro-2-methylpyrimidin-4-ol, 6-chloro-2-methylpyrimidin-4 1h-one, 4 1h-pyrimidinone, 6-chloro-2-methyl, chloromethylpyrimidinol, 4 3h-pyrimidinone, 6-chloro-2-methyl, 4-pyrimidinol, 6-chloro-2-methyl, 6-chloro-2-methyl-4 3h-pyrimidone, 2-methyl-4-hydroxy-6-chloropyrimidine, 4-chloro-6-hydroxy-2-methylpyrimidine
PubChem CID 295975
IUPAC Name 6-chloro-2-methyl-1H-pyrimidin-4-one
SMILES CC1=NC(=O)C=C(N1)Cl

Specifications

Melting Point 207°C to 211°C
Quantity 1 g
Formula Weight 144.56
Percent Purity 97%
Chemical Name or Material 6-Chloro-4-hydroxy-2-methylpyrimidine
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only