accessibility menu, dialog, popup

UserName

CAS RN 17551-52-9

IUPAC Name: 6-chloro-2-methyl-1,4-dihydropyrimidin-4-one
Synonyms: 6-chloro-2-methyl-1H-pyrimidin-4-one
Molecular Weight (g/mol): 144.56
Molecular Formula: C5H5ClN2O
InChi Key: QNANRGHPXMUJQC-UHFFFAOYSA-N
SMILES: CC1=NC(=O)C=C(Cl)N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results