missing translation for 'onlineSavingsMsg'
Learn More

6-chloro-3-nitro-2-picoline, 98%, Thermo Scientific Chemicals

Catalog No. p-3735999
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC360820010 1 g Glass bottle
AC360820050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC360820010 Supplier Thermo Scientific Chemicals Supplier No. 360820010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 22280-60-0 | Molecular Formula: C6H5ClN2O2 | Molecular Weight: 172.5 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 22280-60-0
Molecular Formula C6H5ClN2O2
Molecular Weight (g/mol) 172.5
MDL Number MFCD00234160
InChI Key GHSRMSJVYMITDX-UHFFFAOYSA-N
Synonym 2-chloro-5-nitro-6-methylpyridine, 6-chloro-3-nitro-2-picoline, 2-chloro-5-nitro-6-picoline, 6-chloro-2-methyl-3-nitro-pyridine, 6-chloro-3-nitro-2-methylpyridine, pyridine, 6-chloro-2-methyl-3-nitro, 2-chloro-6-methyl-5-nitropyridine, 2-chloro-5-nitro-6-picoline 2-chloro-6-methyl-5-nitropyridine, pubchem6225, acmc-1cg7s
PubChem CID 253123
IUPAC Name 6-chloro-2-methyl-3-nitropyridine
SMILES CC1=C(C=CC(=N1)Cl)[N+](=O)[O-]

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 54.0°C to 58.0°C
Color Beige to Brown
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 172.5
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 6-chloro-3-nitro-2-picoline
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if inhaled.
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only