missing translation for 'onlineSavingsMsg'
Learn More

6-chloro-2-picoline, 98%, Thermo Scientific™

Chemical Identifiers

CAS 18368-63-3
Molecular Formula C6H6ClN
Molecular Weight (g/mol) 127.57
MDL Number MFCD00006245
InChI Key GXZDYRYYNXYPMQ-UHFFFAOYSA-N
Synonym 6-chloro-2-picoline, 6-chloro-2-methylpyridine, pyridine, 2-chloro-6-methyl, 2-chloro-6-picoline, 2-picoline, 6-chloro, unii-39mx1lq68l, 2-chloro-6-methyl pyridine, 2-chloro-6-methyl-pyridine, 2-chloro-6-methy pyridine
PubChem CID 87601
IUPAC Name 2-chloro-6-methylpyridine
SMILES CC1=CC=CC(Cl)=N1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC152230010
SDS View Documents
Thermo Scientific Chemicals
152230010
1g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC152230100
SDS View Documents
Thermo Scientific Chemicals
152230100
10g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

18368-63-3
127.57
GXZDYRYYNXYPMQ-UHFFFAOYSA-N
87601
CC1=CC=CC(Cl)=N1
C6H6ClN
MFCD00006245
6-chloro-2-picoline, 6-chloro-2-methylpyridine, pyridine, 2-chloro-6-methyl, 2-chloro-6-picoline, 2-picoline, 6-chloro, unii-39mx1lq68l, 2-chloro-6-methyl pyridine, 2-chloro-6-methyl-pyridine, 2-chloro-6-methy pyridine
2-chloro-6-methylpyridine

Specifications

18368-63-3
100.0
64°C to 68°C (10.0mmHg)
Authentic
Glass bottle
1.5260 to 1.528
MFCD00006245
20, I, 84
6-chloro-2-picoline, 6-chloro-2-methylpyridine, pyridine, 2-chloro-6-methyl, 2-chloro-6-picoline, 2-picoline, 6-chloro, unii-39mx1lq68l, 2-chloro-6-methyl pyridine, 2-chloro-6-methyl-pyridine, 2-chloro-6-methy pyridine
CC1=CC=CC(Cl)=N1
127.57
127.57
6-chloro-2-picoline
97.5
1.1600g/mL
73°C
97.5% min. (GC)
C6H6ClN
CH3(C5H3N)Cl
1g
1.16
GXZDYRYYNXYPMQ-UHFFFAOYSA-N
2-chloro-6-methylpyridine
87601
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER

GHS Signal Word: Warning

EINECSNumber : 242-241-6

SDS
Documents

Documents

RUO – Research Use Only