missing translation for 'onlineSavingsMsg'
Learn More

6-Chloro-2-methyl-2H-1,4-benzoxazin-3(4H)-one 98.0+%, TCI America™

Catalog No. C31381G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C31381G 1 g
C31385G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C31381G Supplier TCI America Supplier No. C31381G

Chemical Identifiers

CAS 5791-00-4
Molecular Formula C9H8ClNO2
Molecular Weight (g/mol) 197.618
MDL Number MFCD01744501
InChI Key MGOCBXKZDZRPMK-UHFFFAOYSA-N
PubChem CID 22035
IUPAC Name 6-chloro-2-methyl-4H-1,4-benzoxazin-3-one
SMILES CC1C(=O)NC2=C(O1)C=CC(=C2)Cl

Specifications

Melting Point 174°C
Quantity 1 g
Formula Weight 197.62
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 6-Chloro-2-methyl-2H-1,4-benzoxazin-3(4H)-one
RTECSNumber DM3750000
TSCA No