missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6-Chloro-1-tetralone 96.0+%, TCI America™
$223.68 - $781.03
Chemical Identifiers
| CAS | 26673-31-4 |
|---|---|
| Molecular Formula | C10H9ClO |
| Molecular Weight (g/mol) | 180.63 |
| MDL Number | MFCD04114377 |
| InChI Key | WQKHERPPDYPMNX-UHFFFAOYSA-N |
| Synonym | 6-Chloro-3,4-dihydro-1(2H)-naphthalenone |
| PubChem CID | 12291149 |
| IUPAC Name | 6-chloro-1,2,3,4-tetrahydronaphthalen-1-one |
| SMILES | ClC1=CC2=C(C=C1)C(=O)CCC2 |
Chemical Identifiers
| 26673-31-4 | |
| 180.63 | |
| WQKHERPPDYPMNX-UHFFFAOYSA-N | |
| 12291149 | |
| ClC1=CC2=C(C=C1)C(=O)CCC2 |
| C10H9ClO | |
| MFCD04114377 | |
| 6-Chloro-3,4-dihydro-1(2H)-naphthalenone | |
| 6-chloro-1,2,3,4-tetrahydronaphthalen-1-one |
Specifications
| 26673-31-4 | |
| 150°C | |
| MFCD04114377 | |
| 6-Chloro-3,4-dihydro-1(2H)-naphthalenone | |
| ClC1=CC2=C(C=C1)C(=O)CCC2 | |
| 180.63 | |
| 180.63 | |
| Crystalline Lumps |
| White-Yellow | |
| C10H9ClO | |
| 1 g | |
| WQKHERPPDYPMNX-UHFFFAOYSA-N | |
| 6-chloro-1,2,3,4-tetrahydronaphthalen-1-one | |
| 12291149 | |
| ≥96.0% (GC) | |
| 6-Chloro-1-tetralone |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator