missing translation for 'onlineSavingsMsg'
Learn More

6-Chloro-1-tetralone 96.0+%, TCI America™

Catalog No. C29175G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C29175G 5 g
C29171G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C29175G Supplier TCI America Supplier No. C29175G
Only null left

Chemical Identifiers

CAS 26673-31-4
Molecular Formula C10H9ClO
Molecular Weight (g/mol) 180.63
MDL Number MFCD04114377
InChI Key WQKHERPPDYPMNX-UHFFFAOYSA-N
Synonym 6-Chloro-3,4-dihydro-1(2H)-naphthalenone
PubChem CID 12291149
IUPAC Name 6-chloro-1,2,3,4-tetrahydronaphthalen-1-one
SMILES ClC1=CC2=C(C=C1)C(=O)CCC2

Specifications

Color White-Yellow
Boiling Point 150°C
Quantity 5 g
Formula Weight 180.63
Percent Purity ≥96.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 6-Chloro-1-tetralone
TSCA No
Recommended Storage Refrigerator