missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6-Chloro-1-tetralone 96.0+%, TCI America™
Specifications
Specifications
| Color | White-Yellow |
| Boiling Point | 150°C |
| Quantity | 1 g |
| Formula Weight | 180.63 |
| Percent Purity | ≥96.0% (GC) |
| Physical Form | Crystalline Lumps |
| Chemical Name or Material | 6-Chloro-1-tetralone |
Safety and Handling
| TSCA | No |
| Recommended Storage | Refrigerator |