missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6-Chloro-1-hexyne 97.0+%, TCI America™
Supplier: TCI America C149325ML
Specifications
| 6-Chloro-1-hexyne | |
| Yellow | |
| C6H9Cl | |
| 25 mL | |
| 6-chloro-1-hexyne, 5-hexynechloride, 1-hexyne,6-chloro, 1-hexyne, 6-chloro, 5-hexynyl chloride, 1-chloro-5-hexyne, 6-chloro-hex-1-yne, acmc-1boe0, zukocgmvjuxija-uhfffaoysa | |
| ClCCCCC#C | |
| 116.59 | |
| 116.59 | |
| Liquid |
| 10297-06-0 | |
| 144°C | |
| MFCD00013697 | |
| 1993 | |
| ZUKOCGMVJUXIJA-UHFFFAOYSA-N | |
| 6-chlorohex-1-yne | |
| 272001 | |
| ≥97.0% (GC) |
Chemical Identifiers
| 10297-06-0 | |
| 116.59 | |
| ZUKOCGMVJUXIJA-UHFFFAOYSA-N | |
| 6-chlorohex-1-yne |
| C6H9Cl | |
| MFCD00013697 | |
| 272001 | |
| ClCCCCC#C |
Safety and Handling
TSCA : No