missing translation for 'onlineSavingsMsg'
Learn More

6-Chloro-1-hexyne 97.0+%, TCI America™

Catalog No. p-7125015
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C14935ML 5 mL
C149325ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C14935ML Supplier TCI America Supplier No. C14935ML
Only null left

Chemical Identifiers

CAS 10297-06-0
Molecular Formula C6H9Cl
Molecular Weight (g/mol) 116.59
MDL Number MFCD00013697
InChI Key ZUKOCGMVJUXIJA-UHFFFAOYSA-N
PubChem CID 272001
IUPAC Name 6-chlorohex-1-yne
SMILES ClCCCCC#C

Specifications

Color Yellow
Boiling Point 144°C
Quantity 5 mL
UN Number 1993
Synonym 6-chloro-1-hexyne, 5-hexynechloride, 1-hexyne,6-chloro, 1-hexyne, 6-chloro, 5-hexynyl chloride, 1-chloro-5-hexyne, 6-chloro-hex-1-yne, acmc-1boe0, zukocgmvjuxija-uhfffaoysa
Formula Weight 116.59
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 6-Chloro-1-hexyne