missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6-Chloro-1,2,3,4-tetrahydrocarbazole 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 149°C |
| Color | White-Yellow |
| Quantity | 1 g |
| Formula Weight | 205.69 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 6-Chloro-1,2,3,4-tetrahydrocarbazole |
Safety and Handling
| TSCA | No |