CAS RN 36684-65-8
CAS RN 36684-65-8
IUPAC Name:
6-chloro-2,3,4,9-tetrahydro-1H-carbazole
Synonyms:
3-chloro-6,7,8,9-tetrahydro-5H-carbazole
Molecular Weight (g/mol):
205.69
Molecular Formula:
C12H12ClN
InChi Key:
CNQQPGJXOHSTQR-UHFFFAOYSA-N
SMILES:
ClC1=CC=C2NC3=C(CCCC3)C2=C1
1
–
1
of
1
results
1
6-Chloro-1,2,3,4-tetrahydrocarbazole 98.0+%, TCI America™
CAS: 36684-65-8 Molecular Formula: C12H12ClN Molecular Weight (g/mol): 205.69 MDL Number: MFCD00541331 InChI Key: CNQQPGJXOHSTQR-UHFFFAOYSA-N PubChem CID: 326615 IUPAC Name: 6-chloro-2,3,4,9-tetrahydro-1H-carbazole SMILES: ClC1=CC=C2NC3=C(CCCC3)C2=C1