missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals 6-Carboxyfluorescein, 96%

Catalog No. AC425801000
Change view
Click to view available options
Quantity:
100 mg
500 mg
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC425801000 100 mg Glass bottle
AC425805000 500 mg Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC425801000 Supplier Thermo Scientific Chemicals Supplier No. 425801000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 3301-79-9 | C21H12O7 | 376.32 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3301-79-9
Molecular Formula C21H12O7
Molecular Weight (g/mol) 376.32
MDL Number MFCD00036873
InChI Key BZTDTCNHAFUJOG-UHFFFAOYSA-N
Synonym 6-carboxyfluorescein, 6-fam, carboxyfluorescein, spiro isobenzofuran-1 3h ,9'-9h xanthene-6-carboxylic acid, 3',6'-dihydroxy-3-oxo, 3,6,9-trihydroxyxanthen-9-yl terephthalic acid, 3',6'-dihydroxy-3-oxo-3h-spiro 2-benzofuran-1,9'-xanthene-6-carboxylic acid, 3',6'-dihydroxy-3-oxo-3h-spiro isobenzofuran-1,9'-xanthene-6-carboxylic acid, 5 6-carboxy fluorescein, bidd:gt0504
PubChem CID 76806
ChEBI CHEBI:39073
IUPAC Name 3',6'-dihydroxy-1-oxospiro[2-benzofuran-3,9'-xanthene]-5-carboxylic acid
SMILES C1=CC2=C(C=C1C(=O)O)C3(C4=C(C=C(C=C4)O)OC5=C3C=CC(=C5)O)OC2=O

Specifications

CAS Min % 95.0
CAS Max % 100.0
Melting Point >300.0°C
Color Yellow to Orange
Infrared Spectrum Authentic
Assay Percent Range 95% min. (HPLC)
Packaging Glass bottle
Quantity 100 mg
Formula Weight 376.32
Percent Purity 96%
Physical Form Powder
Chemical Name or Material 6-Carboxyfluorescein
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only