missing translation for 'onlineSavingsMsg'
Learn More

6-Bromoquinoline 95.0+%, TCI America™

Catalog No. p-7119422
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
5 g
Catalog No. Quantity
B20151G 1 g
B20155G 5 g
2 options

Catalog No. B20151G

Supplier: TCI America B20151G

Only null left

Chemical Identifiers

CAS 5332-25-2
Molecular Formula C9H6BrN
Molecular Weight (g/mol) 208.058
MDL Number MFCD00024023
InChI Key IFIHYLCUKYCKRH-UHFFFAOYSA-N
Synonym quinoline, 6-bromo, 6-bromo-quinoline, 6-bromo quinoline, 6-bromchinolin, 6-bromchinolin;, 6-br-quinoline, 6-bromooquinoline, acmc-20aits, pubchem2374, 6-bromoquinoline
PubChem CID 79243
IUPAC Name 6-bromoquinoline
SMILES C1=CC2=C(C=CC(=C2)Br)N=C1

Specifications

Color Yellow
Boiling Point 116°C
Quantity 1 g
Formula Weight 208.06
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 6-Bromoquinoline
TSCA Yes
Recommended Storage Refrigerator